This item is now discontinued. Please browse similar items
SHOWA 8115-09 Cut-Resistant Glove, Uncoated; size, large
Representative image only.
SHOWA Mfr # 8115-09 – Item # RK-86483-32
No need to sacrifice tactile sensitivity for cut-resistant protection
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)15
  • MaterialDyneema®
  • SizeLarge
  • ColorGray
  • ComplianceFDA CR21
  • SterileNo
  • DescriptionCut-Resistant Glove, Uncoated; size, large
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
Made of Dyneema®-Spectra™ engineered fibers, this lightweight glove and sleeve offer an ANSI/ ASTM Level IV cut rating. Ideal for handling box cutters, knives, shears, and fabricated and sharp edged metals that require a high level of dexterity — useful in meat and poultry processing. Choose T-Flex Plus® gloves with coated dots for increased dexterity. All are fully washable. Ambidextrous. Approval: CE
Compliance: Made with FDA compliant material for food handling