SHOWA 8814-9 Heat Resistant Glove, 14"L, Size 9
Representative image only.
SHOWA Mfr # 8814-9 – Item # RK-86483-87
High heat resistance up to 500°F (260°C) with superior dexterity
PRODUCT OPTIONS
$25.78CAD / Pair of 1
BUY MORE & SAVE
  • 1-11$25.78CAD / Pair of 1
  • 12+$23.20CAD / Pair of 1
SPECS & DESCRIPTION
Specifications & Description
  • Size9
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • SterileNo
  • DescriptionHeat Resistant Glove, 14\"L, Size 9
Conformite Europeenne
More About this Item
Ideal for use with autoclaves, food service, inspection, and more! Thermal-protective gloves feature a non-woven liner for excellent flexibility and fit. Designed to withstand heat exposure and maintain superior tactile dexterity, even after multiple washings. Textured gloves provide an excellent grip. The black color doesn't show dirt and grime extending its wearable life.