SHOWA 707-09 Nitrile Glove, Size 9, Package of 12 pair
Representative image only.
SHOWA Mfr # 707D-09 – Item # RK-86483-15
Has the same feel as gloves half as thick
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
PRODUCT OPTIONS
$24.31CAD / Pkg of 12
BUY MORE & SAVE
  • 1-11$24.31CAD / Pkg of 12
  • 12+$21.88CAD / Pkg of 12
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)9
  • MaterialNitrile
  • Size9
  • Length (in)12
  • Length (cm)30.5
  • ColorBlue
  • GripTractor-tread
  • SterileNo
  • DescriptionNitrile Glove, Size 9, Package of 12 pair
Conformite Europeenne
Key Features
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
More About this Item
These 100% nitrile gloves are made with a low modulus hybrid-formulation — meaning that this 9-mil thick glove has the same tactile feel as the thinner Best N-Dex®. Additionally, the softer nitrile formula causes less stress on your hand. Tractor-tread pattern helps with grip. Hand-specific packaging for less waste. Unlined.