Seward BA6041 Stomacher® 400 Circulator Standard Bags, Square Bottom; 500/CS
Representative image only.
Seward Mfr # BA6041 – Item # RK-36850-97
Eliminates the risk of crosss contamination
  • 1 year warranty
  • Manufactured from high-grade, food quality polythene
  • Minimum 65 micron film thickness provides high-impact strength
  • All Seward Stomacher Bags are manufactured from virgin LDPE polymer. The only seal is at the base.
  • Compatible with all paddle lab blender, homogenizers.
  • All Seward Stomacher Bags are gamma irradiated at 25 to 50kGy to guarantee sterility.
  • 1 year warranty
PRODUCT OPTIONS
$211.06CAD / Case of 500
Often bought with...
SPECS & DESCRIPTION
Specifications & Description
  • Bag TypePaddle Blender Bags
  • Bag Capacity (mL)400
  • Bag Capacity (L)0.4
  • DescriptionStomacher bags for Stomacher 400C; square bottom; 500/cs
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
  • Manufactured from high-grade, food quality polythene
  • Minimum 65 micron film thickness provides high-impact strength
  • All Seward Stomacher Bags are manufactured from virgin LDPE polymer. The only seal is at the base.
  • Compatible with all paddle lab blender, homogenizers.
  • All Seward Stomacher Bags are gamma irradiated at 25 to 50kGy to guarantee sterility.
  • 1 year warranty
More About this Item
Choose from a variety of bags to use with your lab blender. Blending bags are made from high quality food grade nontoxic virgin LDPE granule, extruded to 65µm film thickness. All blender bags are irradiated sterile. A certificate or irridiation is available upon request.
The Stomacher bag range has been designed to fulfil all applications from sample blending, straining and storage, including bags designed for extensive blending (processes requiring more than 15 minutes on the machine) and small samples for life science applications.