This item is now discontinued. We recommend the following as suggested replacements: RK-81780-27

SciMatCo SC8080 Flammable Storage Cabinet, 24 gallon capacity, manual-latching door

Representative image only.
SciMatCo Mfr # SC8080 – Item # RK-81781-30
Wooden cabinets provide excellent thermal insulation in case of fire
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • MaterialWood
  • Capacity (Liters)90.8
  • Capacity (Gal)24
  • Number of Shelves1
  • Width (in)30
  • Height (in)32
  • Depth (InchShort)18 1/2
  • Width (cm)76.2
  • Height (cm)81.3
  • Depth (cm)47
  • ColorYellow
  • Self-ClosingNo
  • ComplianceOSHA, NFPA, UL Listed
  • Shipping Weight (lbs)115
  • DescriptionFlammable Storage Cabinet, 24 gallon capacity, manual-latching door
  • Warranty1 Year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

Use this lockable, wooden cabinet to conveniently store flammable liquids. Constructed of 1" (2.5 cm) thick exterior grade plywood; finished with yellow epoxy paint inside and out. Specially designed floor serves as a liquid-tight trough to retain spills. Locking door features continuous piano-type hinges, and a 8" x 8" (20.3 x 20.3 cm) "Flammable" hazard label. Contains one variable position shelf. Meets all applicable OSHA and NFPA standards for flammable safety cabinets. UL Listed.

Meet all applicable OSHA and NFPA standards for flammable safety cabinets. UL Listed

Compliance: OSHA, NFPA

Approvals UL Listed

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat