This item is now discontinued. Please browse similar items

Scientific Industries Genie SI-1400 Incubator-, Incubating Rotating Shaker; 120V

Representative image only.
Scientific Industries Mfr # SI-1400 – Item # RK-51603-01
Incubate, shake, rock, and rotate all in one compact unit.
  • Magnetic platform can be set to rock or rotate almost any container
  • Offers precise temperature control of ±0.2°C
  • Variety of accessories make this shaker versatile for any lab
  • 2 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Min Speed (rpm)3
  • Max Speed (rpm)35
  • Max Load (lbs)10
  • Width (in)22
  • Height (in)14.5
  • Depth (in)15.75
  • Width (cm)55.88
  • Height (cm)36.83
  • Depth (cm)40.005
  • Max Operating Temperature (° F)167
  • Max Operating Temperature (° C)75
  • Temperature Accuracy±0.2°C
  • Temperature Uniformity±0.5°C
  • Power (VAC)120
  • Power (Hz)60
  • DescriptionIncubator-, Incubating Rotating Shaker; 120V
  • Warranty2 year
Two year warrantyConformite Europeenne

Key Features

  • Magnetic platform can be set to rock or rotate almost any container
  • Offers precise temperature control of ±0.2°C
  • Variety of accessories make this shaker versatile for any lab
  • 2 year warranty

More About this Item

Use this 3-in-1 unit to perform assays, washings, and hybridizations more efficiently. Air within the chamber is continuously and gently recirculated to ensure consistent and uniform temperature from 28 to 75°C. Digital control makes it simple to set rotating/rocking speed and desired temperature which are individually displayed for reproducible conditions.

A shatterproof window allows viewing of samples without disruption of chamber environment. Magnetic stainless steel platform and clip plate system hold virtually any container at any angle.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat