Scienceware H13201-0001 Biohazard Autoclave Gloves, 232C max, 18.5" L

Representative image only.
Scienceware Mfr # H13201-0001 – Item # RK-06496-42
Heat resistant to 450°F (232°C)
  • Heat resistant to 450°F (232°C)

PRODUCT OPTIONS

$92.82CAD / Pair of 1

BUY MORE & SAVE

  • 1-11$92.82CAD / Pair of 1
  • 12+$83.54CAD / Pair of 1

SPECS & DESCRIPTION

Specifications & Description

  • MaterialCotton
  • SizeOne size fits all
  • Length (in)18 1/2
  • Length (cm)47
  • ColorOrange
  • SterileNo
  • DescriptionBiohazard Autoclave Gloves, 232C max, 18.5\" L
Conformite Europeenne

Key Features

  • Heat resistant to 450°F (232°C)

More About this Item

Use these soft, pliant, and all-cotton terry cloth gloves to handle hot objects fresh from the autoclave or oven. Excellent for use in any environment where heat protection is required. Bright orange for increased visibility. Machine wash, tumble dry.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat