This item is now discontinued. Please browse similar items
Scienceware F37739-0002 Heavy-Duty Steel Reinforced PE Policeman, 8" L; 6/Pk
Representative image only.
Scienceware Mfr # F37739-0002 – Item # RK-06262-50
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyethylene
  • SterileYes
  • DescriptionHeavy-Duty Steel Reinforced PE Policeman, 8\" L; 6/Pk
More About this Item
Policeman is reinforced with a 1⁄4" (6 mm) steel rod to provide added strength and enclosed in polyethylene for superior chemical resistance. Tapered, flat end measures 1" (2.5 cm) L x 1" (2.5 cm) W. Temperature range is –58 to 176°F (–50 to 80°C).