This item is now discontinued. We recommend the following as suggested replacements: RK-06264-05
Scienceware F36727-0000 Polypropylene sampling spoon assortment pack
Representative image only.
Scienceware Mfr # F36727-0000 – Item # RK-06263-55
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • SterileNo
  • DescriptionPolypropylene sampling spoon assortment pack
More About this Item
Sampling spoons made of talc-filled polypropylene are autoclavable at 250° (121°C) to maintain sample integrity. Select from various capacities to meet your laboratory needs.