This item is now discontinued. Please browse similar items
Scienceware F18905-0051 Centrifuge Tube Ice Rack Cover, polypropylene
Representative image only.
Scienceware Mfr # F18905-0051 – Item # RK-06274-06
For temperature stabilized benchtop ice storage
SPECS & DESCRIPTION
Specifications & Description
  • Min Tube Size (mL)15
  • Max Tube Size (mL)50
  • MaterialPolypropylene(PP)
  • DescriptionCentrifuge Tube Ice Rack Cover, polypropylene
More About this Item
This heavy duty, all polypropylene rack provides efficient and extended cooling of twenty-eight 15 ml or 50 ml centrifuge tubes in a chilled bath. The deep bottom tray holds approximately 2 liters of ice flakes and water so that tubes can rest on the bottom of the tray for maximum stability and exposure to the bath. A flat perimeter rim provides space for labeling tape. The rack is steam autoclavable at 121°C (250°F) and stackable with identical racks when full or empty. To further protect tubes during transport or storage while also maintaining stackability, an optional polypropylene cover is available (order separately).