This item is now discontinued. Please browse similar items
Scienceware F18848-4002 Polyurethane Foam Ice Bucket, Black, 4L Capacity
Representative image only.
Scienceware Mfr # F18848-4002 – Item # RK-06274-21
Store up to 4 liters of ice on laboratory benchtops for extended periods of time
  • Excellent temperature insulation prevents ice from melting for hours
  • Tapered design allows convenient stacking of buckets even when full
  • Rigid polyurethane foam is waterproof and will not rust, leak, or sweat
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (cu ft)0.141
  • DescriptionPolyurethane Foam Ice Bucket, Black, 4L Capacity
Key Features
  • Excellent temperature insulation prevents ice from melting for hours
  • Tapered design allows convenient stacking of buckets even when full
  • Rigid polyurethane foam is waterproof and will not rust, leak, or sweat
More About this Item
This durable ice bucket keeps bottles, test tubes, and temperature-sensitive samples cool for hours. Bucket is inert to acetone, liquid nitrogen, dry ice, and salt. Knob in the center of lid allows easy removal and convenient stacking of buckets for storage.