This item is now discontinued. Please browse similar items
Scienceware Aero Eyewash; Buffered, pH Neutral 16oz, 3 bottles/pack
Representative image only.
Scienceware Mfr # 248820016 – Item # RK-12345-70
Provides immediate first aid for eye injuries
  • Effectively rinses impurities and irritants from your eyes
  • Neutralizes acids and alkalis that may enter eyes
  • Offers four year shelf life from date of manufacture
  • Meets all FDA requirements including buffered pH solution and preservative as per 21 CFR part 349
  • Unique eye-opener on 16 oz bottles keeps used fluids away from eyes
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • ComplianceANSI Z358.1-2009
  • DescriptionEyewash; Buffered, pH Neutral 16oz, 3 bottles/pack
  • Warranty3 month
3 Month Warranty
Key Features
  • Effectively rinses impurities and irritants from your eyes
  • Neutralizes acids and alkalis that may enter eyes
  • Offers four year shelf life from date of manufacture
  • Meets all FDA requirements including buffered pH solution and preservative as per 21 CFR part 349
  • Unique eye-opener on 16 oz bottles keeps used fluids away from eyes
  • 3 month warranty
More About this Item
The Aero Emergency Eye Wash combines sterile saline with buffered, pH solution to provide fast, safe, and effective first aid for eye injuries. The all-in-one solution quickly neutralizes acid and corrosive chemicals, returning eyes to normal 7.4 pH levels. Ideal for use in laboratory, classroom, work site, and public facilities.
Compliance: ANSI Z358.1-2009
Available for sale in the United States only.