Sartorius Tacta® LH-729120 Eight-Channel, Adjustable-Volume Pipette; 0.5 to 10 µL
Representative image only.
Sartorius Mfr # LH-729120 – Item # RK-24505-41
Superb comfort with effortless volume adjustment and locking
  • Achieve reliable results with easy calibration adjustment for various fluid types
  • Hold comfortably with the ergonomically designed handle
  • Autoclave fully without disassembly for quick and easy cleaning
  • 2 year warranty
PRODUCT OPTIONS
$1,915.41CAD / Each
$596.72 - $704.28CAD / Each
for an additional
$596.72CAD / Each
for an additional
$704.28CAD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Volume (µL)10
  • AutoclavableYes
  • Min Volume (µL)0.5
  • Min Volume (mL)0.0005
  • Max Volume (mL)0.01
  • Increments (µL)0.01
  • Accuracy (%)±4.0% to ±1.5%
  • Precision (%)±4.0% to ±1.0%
  • Number Of Channels8
  • Tip EjectorYes
  • Color CodeGray
  • DescriptionEight-Channel, Adjustable-Volume Pipette; 0.5 to 10 µL
  • Warranty2 year
Two year warrantyConformite Europeenne
Key Features
  • Achieve reliable results with easy calibration adjustment for various fluid types
  • Hold comfortably with the ergonomically designed handle
  • Autoclave fully without disassembly for quick and easy cleaning
  • 2 year warranty
More About this Item
This premium mechanical pipette has the Optiload feature in both single- and multichannel models for light tip loading with perfect tip sealing. The Optiject soft-tip ejection feature detaches the tip in a smooth, controlled manner. Optilock’s dual-function volume lock offers protection against accidental volume changes.
Use the easy-to-read, four-digit volume display to set exact volumes. Different volume options are color-coded for quick selection of corresponding pipette tips. Safe-Cone filters (included in models above 10 μL) protect against contamination. Pipettors are made of materials which have high chemical and UV resistance to ensure a long life span.