This item is now discontinued. Please browse similar items
Sample Spatula with Stainless Steel Blade and Wooden Handle
Representative image only.
Item # RK-33687-34
Large blade makes sample collecting easy
  • Easy to grip wooden handle
  • Quickly covers a wide area
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel Blade, Wooden Handle
  • Length (in)11.6
  • Width (in)1.25
  • Length (cm)29.5
  • Width (cm)3.2
  • DescriptionSample Spatula, stainless steel blade w/ wood handle
  • Warranty1 year
1 Year Warranty
Key Features
  • Easy to grip wooden handle
  • Quickly covers a wide area
  • 1 year warranty
More About this Item
This durable spatula has a large blade which makes collecting samples easy. Use to collect solid surface samples or to remove the first layer and collect samples that are beneath the surface. Blade is made of stainless steel and includes a wooden handle for a comfortable grip. Spatula, including handle, measures 295 mm L x 32 mm W (11.6"L x 1.25"W).