This item is now discontinued. Please browse similar items
Salisbury by Honeywell SKCA8-M Arc Flash Protective Coverall Kits, 8 cal/cm2, medium
Representative image only.
Salisbury by Honeywell Mfr # SKCA8-M – Item # RK-86436-60
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • SizeMedium
  • ComplianceHazard Risk Category 2
  • DescriptionArc Flash Protective Coverall Kits, 8 cal/cm2, medium
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Take these kits wherever you need arc flash protection, kits come with a convenient carrying bag. Include coveralls, safety glasses, hood, and helmet with face shield
Coverall features
* Made with Indura Ultra Soft® (88% cotton) for comfort and breathability
* Cuff openings and front closure are hook & pile
* 30" (762 mm) inseam with expansion back
* Meet current ASTM F1506 and NFPA 70E standards
* CE 0120 certified
Hood features
* Head and neck protection from arc flash dangers
* Made of two layers of rib knit material with an elastic face opening
Helmet with face shield features
* Face shield has 70% visual light transmission (VLT) — best in the industry!
* 7.5" x 20" viewing area
* Absorbs 99.9% of harmful UV radiation
* Tested to ASTM F2178 specifications