This item is now discontinued. Please browse similar items

Salisbury by Honeywell SK40LT-2 Arc Flash Lightweight Protective Flash Suit Kit, XXL, 40 cal/cm2; 1/Pk

Representative image only.
Salisbury by Honeywell Mfr # SK40PLT-2XL – Item # RK-86436-87
Protective suits meet NFPA 70E hazard risk category (HRC) 4
  • Made with Indura Ultra Soft® (88% cotton) for comfort and breathability
  • Meets current ASTM F2178 and NFPA 70E standards and the flammability requirements for ASTM F1506
  • Lightweight material is 10% lighter than the standard, making the job more comfortable
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Length (in)32
  • Length (cm)81.28
  • SizeXX-Large
  • ColorGray
  • ComplianceHazard Risk Category 4
  • DescriptionArc Flash Lightweight Protective Flash Suit Kit, XXL, 40 cal/cm2; 1/Pk
  • Warranty1 year
1 Year Warranty

Key Features

  • Made with Indura Ultra Soft® (88% cotton) for comfort and breathability
  • Meets current ASTM F2178 and NFPA 70E standards and the flammability requirements for ASTM F1506
  • Lightweight material is 10% lighter than the standard, making the job more comfortable
  • 1 year warranty

More About this Item

These kits meet NFPA 70E hazard risk category (HRC) 4. All are rated to 40 cal/cm2. Kit includes an arc flash coat, bib overalls, Pro-Hood™, hard hat, safety glasses, and carrying bag.
Pro-Wear™ Flash coat and bib overall features
* 40 cal/cm2 Arc Thermal Protective Value (ATPV)
* Sewn with Nomex® thread
* Meets current ASTM F1506 and NFPA 70E standards
* CE 0120 certified
* Jackets feature 32" length, Nomex® wristlets, and hook & pile storm closure
* Bib overalls feature 30" inseam, integrated heavy duty suspenders, and hook & pile waist straps
Prohood™ features
* Made from arc flash resistant Indura Ultra Soft®
* Sewn with Nomex® thread
* Arc rated 10" x 20" anti-fogging lens
* CE 0120 certified

Lightweight material is 10% lighter than the standard material making your job more comfortable.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat