This item is now discontinued. Please browse similar items
Salisbury by Honeywell GK214RB12 Insulated Glove Kit, Class 2, Black/Red, 14"L, Size 12; One Pair
Representative image only.
Salisbury by Honeywell Mfr # GK214RB12 – Item # RK-86435-29
Comply with NFPA 70E standard while protecting employees from dangerous electrical injuries
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRubber
  • Size12
  • Length (in)14
  • Length (cm)35
  • ColorBlack
  • ComplianceClass 2
  • SterileNo
  • DescriptionInsulated Glove Kit, Class 2, Black/Red, 14\"L, Size 12; One Pair
More About this Item
Get everything you need to protect yourself and keep your insulating rubber gloves in top working condition. Kits include a pair of insulating rubber gloves; the appropriate leather protector that protects the glove from cuts, abrasions, and punctures; and a storage bag which keeps the glove flat, preventing folds and creases that strain the rubber, shortening the life of the glove.
Note: In accordance with NFPA 70E, anything over 50 volts requires insulating rubber gloves.