Salisbury by Honeywell GK011R12 Insulated Glove Kit, Class 0, Red, 11"L, Size 12; One Pair
Representative image only.
Salisbury by Honeywell Mfr # GK011R/12 – Item # RK-86435-14
Comply with NFPA 70E standard while protecting employees from dangerous electrical injuries
PRODUCT OPTIONS
$339.25CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRubber
  • Size12
  • Length (in)11
  • Length (cm)27.9
  • ColorRed
  • ComplianceClass 0
  • SterileNo
  • DescriptionInsulated Glove Kit, Class 0, Red, 11\"L, Size 12; One Pair
More About this Item
Get everything you need to protect yourself and keep your insulating rubber gloves in top working condition. Kits include a pair of insulating rubber gloves; the appropriate leather protector that protects the glove from cuts, abrasions, and punctures; and a storage bag which keeps the glove flat, preventing folds and creases that strain the rubber, shortening the life of the glove.
Note: In accordance with NFPA 70E, anything over 50 volts requires insulating rubber gloves.