This item is now discontinued. Please browse similar items
Salisbury by Honeywell FH40GY-C Air FLow Hood System, 40 cal/cm2
Representative image only.
Salisbury by Honeywell Mfr # FH40GY-C – Item # RK-86436-90
Arc flash protection and cooling in a comfortable, lightweight unit
  • Arc flash protection hood with an integrated cooling system
  • Provides up to 80 cfm of airflow throughout the breathing zone to help cool your workers
  • Ideal in warm conditions and confined spaces with poor air circulation
  • 40 cal/cm2 Arc Thermal Protective Value (ATPV)
  • Powered by eight AA batteries (not included) with continuous use up to 35 hours
  • System measures 14"L x 7"W (356 x 178 mm) and weighs less than 5 lbs
  • Note: Air is not filtered and the unit is not considered a respirator
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAir FLow Hood System, 40 cal/cm2
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Arc flash protection hood with an integrated cooling system
  • Provides up to 80 cfm of airflow throughout the breathing zone to help cool your workers
  • Ideal in warm conditions and confined spaces with poor air circulation
  • 40 cal/cm2 Arc Thermal Protective Value (ATPV)
  • Powered by eight AA batteries (not included) with continuous use up to 35 hours
  • System measures 14"L x 7"W (356 x 178 mm) and weighs less than 5 lbs
  • Note: Air is not filtered and the unit is not considered a respirator