This item is now discontinued. Please browse similar items
Salisbury by Honeywell E0011B12 Insulated Glove, Class 00, Black, 11" length, size 12
Representative image only.
Salisbury by Honeywell Mfr # E0011B12 – Item # RK-86432-19
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRubber
  • Size12
  • Length (in)11
  • Length (cm)27.9
  • ColorBlack
  • ComplianceClass 00
  • SterileNo
  • DescriptionInsulated Glove, Class 00, Black, 11\" length, size 12
More About this Item
Protect yourself when working with switchboards, power cabinets, or anytime you are working on energized circuits. Combining high dielectric and physical strength, flexibility and durability, gloves provide superior performance meeting and exceeding the requirements of current ASTM D120 specifications and NFPA 70E standards.
* Made of Type I natural rubber
* Meet or exceed ASTM D120 and IEC EN60903
* Available in different colors to allow easy inspection of a glove rotation program
* Glove flexibility makes working with small parts easy
* Glove Inflator (sold separately) simplifies inspection of gloves before each use