This item is now discontinued. Please browse similar items
Salisbury by Honeywell AS1000HAT Arc Flash Protective Face Shield with Helmet
Representative image only.
Salisbury by Honeywell Mfr # AS1000HAT – Item # RK-86436-14
Tested to ASTM F2178 specifications
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • ColorOrange
  • DescriptionArc Flash Protective Face Shield with Helmet
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
* 10 cal/cm2 Arc Thermal Protective Value (ATPV) rating when used with the included chin cup; 8 cal/cm2 without the chin cup
* 7.5" x 20" viewing area
* Extra light tint
* Absorbs >99.9% of harmful UV radiation
* Resistant to fogging
* Tested to ASTM F2178 specifications