Rubberfab TR-PW Torque Tee Sanitary Clamp Securing Device Palm Wrench, 3/8"
Representative image only.
Rubberfab Mfr # TR-PW – Item # RK-23508-06
Ensure the security of sanitary connections with preset torque output
  • Works with any sanitary clamp wing nut and reaches easily into tight or awkward spaces
  • Choose the appropriate torque output per the clamp manufacturer's specifications
  • Audible click alerts you when the torque has been reached
  • Ratcheting mechanism disengages for easy clamp removal and is sealed within handle for durability
  • "T" shape creates effective leverage for fast, easy one-handed tightening
PRODUCT OPTIONS
$77.35CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Drive (in)0.375
  • DescriptionTorque Tee Sanitary Clamp Securing Device Palm Wrench, 3/8"
Key Features
  • Works with any sanitary clamp wing nut and reaches easily into tight or awkward spaces
  • Choose the appropriate torque output per the clamp manufacturer's specifications
  • Audible click alerts you when the torque has been reached
  • Ratcheting mechanism disengages for easy clamp removal and is sealed within handle for durability
  • "T" shape creates effective leverage for fast, easy one-handed tightening
More About this Item
Use the Torque Tee to install and remove sanitary clamps safely and securely, even in hard-to-reach spaces. The shape of the handle provides plenty of leverage to reach the factory preset torque. Just listen for the audible click to know the clamp wing nut is properly tightened. If you need to loosen or remove the clamp, simply disengage the ratcheting mechanism to lock the handle.