Dynalon Round Dipping Basket, PP for Crock 06724-40
Representative image only.
Dynalon Mfr # 167174-30 – Item # RK-06717-30
  • 1 year warranty
PRODUCT OPTIONS
$540.73CAD / Each
BUY MORE & SAVE
  • 1-5$540.73CAD / Each
  • 6-29$486.66CAD / Each
  • 30-119$459.62CAD / Each
  • 120+$432.58CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene
  • Overall Height (in)10
  • Overall Height (cm)25.4
  • Inside Diameter (ID) (in)9
  • Inside Diameter (ID) (cm)22.9
  • DescriptionRound Dipping Basket, PP for Crock 06724-40
  • Warranty1 year
1 Year WarrantyNSF international certified
Key Features
  • 1 year warranty
More About this Item
These polypropylene (PP) baskets withstand temperatures to 250°>F (121°>C). Sides and bottom have 1/8'' perforations. Reinforced top and bottom rings give added strength. Swing-type handle. Use with 06724-series Heavy Crocks.