This item is now discontinued. Please browse similar items
Rite in the Rain 93B All-Weather Pen; Blue Ink
Representative image only.
Rite in the Rain Mfr # 93B – Item # RK-06570-42
Write through water, grease, and mud—without clumping or smearing!
  • Writes in temperatures from -30 to 250°F (-34 to 121°C),
  • Use under water up to 35 ft (10 m)
  • Write at any angle—including upside down
  • Common color selection to meet GDP requirements—blue ink
SPECS & DESCRIPTION
Specifications & Description
  • ColorBlue
  • Length (in)5.375
  • DescriptionAll-Weather Pen, Blue Ink
Key Features
  • Writes in temperatures from -30 to 250°F (-34 to 121°C),
  • Use under water up to 35 ft (10 m)
  • Write at any angle—including upside down
  • Common color selection to meet GDP requirements—blue ink
More About this Item
These All-Weather Pens will writes on paper in any conditions and can even be used upside down. Rubberized grip makes the pen easy to hold—even in the wettest conditions. Pressurized cartridge means the ink won't leak, evaporate, or blow up in your pocket.