Rex Gauge TB-1-D Test Block for Durometers. Type D, Single Block, Color Black
Representative image only.
Rex Gauge Mfr # TD-1-D – Item # RK-59792-52
Optimize performance of your Rex Gauge durometer kit test block
  • Keep your durometer kit test block running properly with these accessories
  • 1 year warranty
PRODUCT OPTIONS
$130.98CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionShore D Test Block for Durometers
  • Warranty1 year
1 Year Warranty
Key Features
  • Keep your durometer kit test block running properly with these accessories
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your Rex Gauge durometer kit test block.
This type D durometer tests the hardness of hard rubber and plastics. To obtain accurate readings using a durometer, one must develop a proper "feel" for the gauge. Rex Gauge's test block for shore D durometers provides you the ability to develop the proper "feel". While not to be used for calibration purposes, this test block enables you to perform a quick check for the proper operation of the durometer.