This item is now discontinued. We recommend the following as suggested replacements: RK-06736-09
Bel-Art Reversible 96-Well Microcentrifuge Tube Rack, Blue
Representative image only.
Bel-Art Mfr # F18845-0029 – Item # RK-06736-08
Withstand temperatures down to -80 °C (-112 °F)
  • Autoclavable
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Tubes Held96
  • Length (in)8 3/8
  • Width (in)4 1/2
  • Height (in)1 1/16
  • MaterialPolypropylene(PP)
  • ColorBlue
  • DescriptionReversible 96-well microcentrifuge tube rack, blue 06736-08
  • Warranty3 month
3 Month Warranty
Key Features
  • Autoclavable
  • 3 month warranty
More About this Item
One side of this polypropylene (PP) racks is designed to hold 0.5mL microcentrifuge tubes; the other side holds 1.5 to 2.0mL tubes. Withstand temperatures down to -80°C (-112°F).