This item is now discontinued. Please browse similar items
Retsch 32.935.0008 Cyclone w/ Collector and Holder for Air-Jet Sieving Machine
Representative image only.
Retsch Mfr # 32.935.0008 – Item # RK-59996-38
Ideal for optimal use of your Air-Jet Sieving Machine
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCyclone w/ Collector and Holder for Air-Jet Sieving Machine
More About this Item
Sieving machine requires a vacuum cleaner to create the suction power, air-jet sieving machine, and 8" diameter test sieve, all sold separately. Other accessories are ideal for optimal perfomance of the sieving machine as well as maintenance. Air sieving machine is available as part number 59996-30.
Exhaust the sieved material through the vacuum, or use the optional cyclone to capture and save the material.