ResinTech Recirculating Dispensing Gun for CLiR 3000 High Purity Water System
Representative image only.
ResinTech Mfr # CLA-3000-DG – Item # RK-99285-24
Accessories, options, and consumables for CLïR Lab Water Systems
  • Made in the USA!
  • Ideal for dispensing water from Type I high purity water systems
  • 1 year warranty
PRODUCT OPTIONS
$1,894.78CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRecirculating Dispensing Gun with 0.2 micron filter
  • Warranty1 year
1 Year Warranty
Key Features
  • Made in the USA!
  • Ideal for dispensing water from Type I high purity water systems
  • 1 year warranty
More About this Item
Choose from a variety of options and accessories for your ResinTech high-purity water system.