This item is now discontinued. Please browse similar items
Red Lion ZMD0250B Miniature Length Sensor Quad Output 250 PPR
Representative image only.
Red Lion Mfr # ZMD0250B – Item # RK-68493-52
Spring-loaded torsion arm for quick installation
  • Compact size
  • Quadrature output
  • Built-in spring tensioning
  • Vertical, horizontal, or upside-down mounting for quick installation
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Supply Voltage (VDC)28
  • Min Supply Voltage (VDC)4.75
  • Actuator Type250 PPR
  • Operating Frequency200 KHz max
  • Max Operating Temperature (° F)185
  • Min Operating Temperature (° F)-4
  • Max Operating Temperature (° C)85
  • Min Operating Temperature (° C)-20
  • Cable Length (ft)6.5
  • Cable Length (m)1.9812
  • DescriptionMiniature Length Sensor Quad Output 250 PPR
  • Warranty2 Years
Two year warranty
Key Features
  • Compact size
  • Quadrature output
  • Built-in spring tensioning
  • Vertical, horizontal, or upside-down mounting for quick installation
  • 2 year warranty
More About this Item
Designed for light to medium duty sensing applications, the Miniature Length Sensor, model ZMD, is compact in size and easy-to-use. It features a built-in spring-loaded torsion arm that provides a simple-to adjust torsion load, allowing the unit to be mounted in almost any orientation, including upside down.
Using a 6" or 200 mm wheel, the ZMD can be used on almost any surface, while operating at speeds up to 3000 feet per minute. The housing is a durable, conductive composite material that will eliminate static build up. Whether you need to measure speed, position, or distances, the Model ZMD is the ideal solution.