This item is now discontinued. Please browse similar items
Red Lion ZBG01002 Single Channel General Service 6-Pin MS Connector 100PPR
Representative image only.
Red Lion Mfr # ZBG01002 – Item # RK-68493-44
High pulse per revolution for accuracy
  • Current sink outputs
  • High pulse per revolution (PPR) rates
  • Up to 1200 PPR for fine, high-resolution counting or precision speed measurement from slow shaft speeds
  • Quadrature output for position measurement, bi-directional counting and systems with backlash counting requirements.
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Supply Voltage (VDC)28
  • Min Supply Voltage (VDC)4.75
  • Output Connector Type6-pin MS style connector
  • Actuator Type100 PPR
  • Operating Frequency20 KHz max
  • Max Operating Temperature (° F)212
  • Min Operating Temperature (° F)32
  • Max Operating Temperature (° C)100
  • Min Operating Temperature (° C)0
  • DescriptionSingle Channel General Service 6-Pin MS Connector 100PPR
  • Warranty2 Years
Two year warranty
Key Features
  • Current sink outputs
  • High pulse per revolution (PPR) rates
  • Up to 1200 PPR for fine, high-resolution counting or precision speed measurement from slow shaft speeds
  • Quadrature output for position measurement, bi-directional counting and systems with backlash counting requirements.
  • 2 year warranty
More About this Item
The units contain an L.E.D. light source and a photo sensor that scans a shaft mounted, slotted disc. An internal pulse-shaping amplifier circuit delivers a rectangular pulse signal from the current sinking output in response to the passing slots as it rotates. They can be direct-coupled to a machine shaft by means of a flexible-bellows, spring, or rubber sleeve type coupling that allows for axial and radial misalignment. They can also be coupled with light instrument timing-belts. Timing-belt drives also allow convenient gear-up or gear-down speed ratio changes that can be useful for obtaining non-standard PPR rates.