This item is now discontinued. Please browse similar items
Red Lion DSPSX000 Operator Interface and Protocol Converter
Representative image only.
Red Lion Mfr # DSPSX000 – Item # RK-68515-99
A nexus for industrial data collection and management
  • Converts numerous protocols simultaneously
  • Over 200 built-in drivers allow easy data mapping to PLCs, PCs, drives and SCADA systems
  • Supports up to nine protocols simultaneously (with expansion card)
  • DIN rail mounting
  • Operating termperature 32 to 122°F (0 to 50°C)
  • Comes with standard web server, data logging (via CSV file format) and alarm e_mail/text messaging
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)5.9062
  • Width (in)8.0312
  • Depth (InchShort)1.8906
  • DescriptionOperator Interface and Protocol Converter
  • Warranty2 Years
Two year warranty
Key Features
  • Converts numerous protocols simultaneously
  • Over 200 built-in drivers allow easy data mapping to PLCs, PCs, drives and SCADA systems
  • Supports up to nine protocols simultaneously (with expansion card)
  • DIN rail mounting
  • Operating termperature 32 to 122°F (0 to 50°C)
  • Comes with standard web server, data logging (via CSV file format) and alarm e_mail/text messaging
  • 2 year warranty
More About this Item
The Data Station Plus is designed to act as a nexus for industrial data collection and management. With three built-in serial ports and a 10 Base-T/100 Base-TX Ethernet port, the unit performs protocol conversion, allowing disparate devices to communicate seamlessly with one another. The Data Station Plus functions both as a data management platform and as a PFM host.
The Ethernet port supports up to four protocols simultaneously so even Ethernet-to-Ethernet protocol conversion may be performed. The CompactFlash card allows new configuration files to be loaded into the Data Station Plus or use to data log critical information. The USB port may be used for fast file downloads, or to mount the Data Station’s CompactFlash card as an external drive to your PC.
The DSPSX version also features e-mailing/text messaging capabilities that allow alarms to be forwarded to any cell phone or network PC, and has a built-in web server, providing worldwide access to data logs and virtual HMI.