Ralston Instruments QTHA-2MS0 Adapter, 1/4" NPT(M) to Male Quick-Test Connection; Stainless Steel
Representative image only.
Ralston Instruments Mfr # QTHA-2MS0 – Item # RK-16110-01
Leak free connections for pressure calibrations
  • 1 year warranty
PRODUCT OPTIONS
$45.09CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure5000 psi (345 bar)
  • Min Pressure0 psi (0 bar)
  • DescriptionAdapter, 1/4\" NPT(M) to Male Quick-Test Connection; Stainless Steel
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
High quality fitting kits and connection hoses are designed to give reliable and leak free connections for pressure calibrations. Since these fittings do not require wrenches or thread sealant, these fitting kits and quick connect hoses reduce calibration time and down time due to calibration of pressure gauges and transmitters.