This item is now discontinued. Please browse similar items
RAE Systems 10-106-10 Gas detection tubes, chlorine, 0.5 to 8 ppm, 10/box
Representative image only.
RAE Systems Mfr # H10-106-10 – Item # RK-86260-71
Sharp, clear color change for unambiguous measurements
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Gas MeasuredChlorine
  • Range0.5 to 8
  • DescriptionGas detection tubes, chlorine, 0.5 to 8 ppm, 10/box
  • Warranty2 year
Two year warranty
Key Features
  • Easy-to-use colorimetric tubes
  • Break off the tube tips at both ends
  • Insert tube into pump (sold separately) and draw sample
  • Read the concentration directly from the printed scale
  • 2 year warranty
More About this Item
Precalibrated, maintenance-free tubes accurately and reproducibly indicate toxic gas concentrations. Box of 10 tubes.