This item is now discontinued. Please browse similar items
Radians DG1-11 Safety Goggles, Clear lens
Representative image only.
Radians Mfr # DG1-11 – Item # RK-86497-50
Flexible enough to be worn as goggles, or as safety glasses
SPECS & DESCRIPTION
Specifications & Description
  • Frame ColorClear
  • Lens ColorClear
  • ComplianceANSI Z87.1-2003
  • DescriptionSafety Goggles, Clear lens
More About this Item
This modern-looking design features a wraparound lens with foam padding that forms a perfect seal. Polycarbonate lens provides impact resistance while seal keeps out dirt and debris. Wear as goggles by using the elastic strap, or switch to the snap-in temples to wear as a pair of safety glasses. Elastic headstrap provides a comfortable, easily adjustable fit. Snap-in temples are ratcheting and telescoping to get the best possible fit.
Compliance: ANSI Z87.1-2003