Qsonica 418 Ultrasonic Cell Disruptor Microtip Probes, 1-16" dia
Representative image only.
Qsonica Mfr # 4417 – Item # RK-04711-89
  • 2 year warranty
PRODUCT OPTIONS
$693.97CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Sample Size (mL)5
  • Tip Diameter (in)0.063
  • Tip Diameter (cm)0.16002
  • DescriptionUltrasonic Cell Disruptor Microtip Probes, 1-16\" dia
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Probes effectively process small samples and attach to the standard 1/2"-dia probe horn. The stepped microtip probe (04711-93) attaches directly to the converter and is ideal for reaching into long, thin tubes.