This item is now discontinued. Please browse similar items
QRP Q125M QualatrileXC Nitrile Gloves, Medium, 12"
Representative image only.
QRP Mfr # Q125M – Item # RK-86409-21
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Thickness (mil)5
  • MaterialNitrile
  • SizeMedium
  • Length (in)12
  • Length (cm)30.5
  • ColorWhite
  • SterileNo
  • DescriptionQualatrileXC Nitrile Gloves, Medium, 12\"
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Powder-free disposable nitrile gloves contain no plasticizers or silicone and are hypoallergenic. Gloves feature a beaded cuff for quick donning and superior snag and puncture resistance. Smooth palm and fingers ensure tactile sensitivity. Low particulate and ion levels make these gloves an excellent choice for use in cleanrooms and laboratories. Formed using left/right molds, these gloves offer superior fit. Left and right gloves are packaged separately and sealed together in bags. Pack of 50 pair.
Compliance: Class 100, ASTM D3578-00, ISO 9002