This item is now discontinued. Please browse similar items
QLA Standard Vessel with Plastic Rim Compatible with Agilent 708/709-DS Baths, 1000 mL, Clear Glass; Serialized
Representative image only.
Mfr # GLA900-70XDS – Item # RK-41306-20
QLA vessels provide more precise and consistent dissolution results
  • Unique serial number is laser marked on plastic rim of each vessel
  • Plastic rims help minimize glass breakage
  • 1000 mL capacity
  • Standard vessel
  • Clear glass
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)1000
  • MaterialGlass
  • Bottle ColorClear
  • Special FeaturesStandard with plastic rim
  • DescriptionStandard Vessel with Plastic Rim Compatible with Agilent 708/709-DS Baths, 1000 mL, Clear Glass; Serialized
Key Features
  • Unique serial number is laser marked on plastic rim of each vessel
  • Plastic rims help minimize glass breakage
  • 1000 mL capacity
  • Standard vessel
  • Clear glass
More About this Item
Glass vessels offer good thermal conductivity and are highly resistance to scratching. Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle.
Not available for sale in the United States.