This item is now discontinued. View the direct replacement: RK-34593-25 or browse similar items

Pyrex 3022-250 Brand 3022 cylinder; 250 mL, pack of 1

Representative image only.
Pyrex Mfr # 3022-250 – Item # RK-34593-05
  • To Contain
  • Single Metric Scale

SPECS & DESCRIPTION

Specifications & Description

  • MaterialGlass
  • Capacity (mL)250
  • Graduation Interval (mL)2
  • Tolerance (mL)±2.0
  • DescriptionBrand 3022 cylinder; 250 mL, pack of 1

Key Features

  • To Contain
  • Single Metric Scale

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat