This item is now discontinued. Please browse similar items
Puma AA1062-blue Air Compressor hose, 1/4 in x 50' PU braid air hose with swivel fitting and grip
Representative image only.
Puma Mfr # AA1062-blue – Item # RK-07123-27
  • Polyurethane, braided air hose is rated to 300 psi working pressure, –32 to 140°F (–35 to 60°C), and available in blue
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Wetted PartsPolyurethane
  • DescriptionAir Compressor hose, 1/4 in x 50' PU braid air hose with swivel fitting and grip
  • Warranty1 year
1 Year Warranty
Key Features
  • Polyurethane, braided air hose is rated to 300 psi working pressure, –32 to 140°F (–35 to 60°C), and available in blue