This item is now discontinued. Please browse similar items
PTFE Round Brush, 7.5" Handle, 2" L x 1" Diameter; 1/Pk
Representative image only.
Item # RK-84552-03
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPTFE
  • Handle Length (in)7.5
  • Length (in)9 1/2
  • DescriptionPTFE Round Brush, 7.5\" Handle, 2\" L x 1\" Diameter; 1/Pk
More About this Item
PTFE brushes are specifically designed for use in high temperature applications. They can withstand temperatures up to 500°F (260°C) and outlast Nylon, plastic and natural Boar bristles. Resistant to many caustic chemicals.