This item is now discontinued. Please browse similar items
PTFE Brush, 5" Handle, 1" L x 3" W; 1/Pk
Representative image only.
Item # RK-84552-01
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPTFE
  • Handle Length (in)5
  • Length (in)6
  • DescriptionPTFE Brush, 5\" Handle, 1\" L x 3\" W; 1/Pk
More About this Item
PTFE brushes are specifically designed for use in high temperature applications. They can withstand temperatures up to 500°F (260°C) and outlast Nylon, plastic and natural Boar bristles. Resistant to many caustic chemicals.