This item is now discontinued. Please browse similar items
Protimeter BLD5365 Surveymaster Moisture Meter
Representative image only.
Protimeter Mfr # BLD5365 – Item # RK-59820-16
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Measurement TypePin
  • Display TypeDigital
  • BatteryOne 9 V battery (included)
  • DescriptionSurverymaster Moisture Meter
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
The Surveymaster is used for measuring themoisture level of non-conductive and porous con-struction materials. It has two modes of operation– Search and Measure. In Search Mode, theSurveymaster is a moisture detector which givesreadings of the moisture condition beneath the sur-face materials. This is ideal for making rapid sur-veys of walls and floors to pinpoint areas of concernthat may require further investigation. This modecan also be used as an alternative to the MeasureMode when it is impractical or undesirable to pushelectrode pins into surfaces. In Measure Mode, theSurveymaster measures the moisture level of thematerial between the two electrodes. It has inte-gral pin electrodes that can be pushed into sur-faces, or it may be used with various auxiliaryprobes including the deep wall probes (supplied) or a hammer electrode (optional).
  • Dual display
  • digital LCD and 60 LEDs
  • Audible tone
  • may be turned on or off by user
  • Auto-Off switch
  • may be set between one and three and minutes