Porta-Lab PSL-2012 Piezo Spark-O-Matic Electronic Spark Lighter
Representative image only.
Mfr # PSL-2012 – Item # RK-36310-60
Sure, safe ignition for your bunsen burner
  • 1 year warranty
PRODUCT OPTIONS
$50.10CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPiezo Spark-O-Matic Electronic Spark Lighter
  • Warranty1 Year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • 1 year warranty
More About this Item
Use this well-constructed, safety spark lighter for all your burner ignition needs. Special spark hood protects the flint. Gas pocket forms beneath the hood, ensuring instant lighting.
Lighter incorporates new larger, renewable flint that produces up to 1500 ignitions..