Pomona Electronics 72902 Test Clip, Set 12 Clips Leads & Rod
Representative image only.
Pomona Electronics Mfr # 72902 – Item # RK-20009-61
Available for sale only in the United States and Canada
  • Kit of 12 micro SMD grabbers in 6 colors
  • 3 month warranty
PRODUCT OPTIONS
$446.42CAD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTest Clip, Set 12 Clips Leads & Rod
  • Warranty3 month
3 Month Warranty
Key Features
  • Kit of 12 micro SMD grabbers in 6 colors
  • 3 month warranty
More About this Item
Test Clips
Micro SMD Grabber Test Clips Kit (PM72902)
Grabbers are designed for easy attachment to SMD chips with pitches down to 0.008". Thin yet durable design permits attachment with close spacing in tight areas. Insulated tip minimizes shorting on adjacent pins. Gold plating for low resistance connections.
Available for sale in the United States and Canada only.