This item is now discontinued. Please browse similar items

Pomona Electronics 3781 Test Clip Patch Cord, 48", Black

Representative image only.
Pomona Electronics Mfr # 3781-48-0 – Item # RK-23031-90
Ideal for providing temporary jumper wires
  • Available in black, 48" length

SPECS & DESCRIPTION

Specifications & Description

  • DescriptionOptional Patch Cord, 48in Black

Key Features

  • Available in black, 48" length

More About this Item

Provides excellent connector to terminals and wire wrap posts.

Available for sale only in the United States and Canada.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat