Dynalon Polypropylene (PP) Paddled Stir Rod, 9-3/4" L; 10/Pk
Representative image only.
Dynalon Mfr # 212455 – Item # RK-06373-00
  • 1 year warranty
$63.36CAD / Pkg of 10
BUY MORE & SAVE
  • 1-5$63.36CAD / Pkg of 10
  • 6-29$57.02CAD / Pkg of 10
  • 30-119$53.86CAD / Pkg of 10
  • 120+$50.69CAD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Handle Length (in)9 3/4
  • Handle Length (cm)24.77
  • Handle Length (ft)0.8125
  • MaterialPolypropylene
  • SterileNo
  • DescriptionPolypropylene (PP) Paddled Stir Rod, 9-3/4\" L; 10/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Use as a stir rod, policeman, or small spatula. Polypropylene (PP) rod features flat and triangular paddles at opposite ends. Max temperature is 266°F (130°C). Autoclavable.