This item is now discontinued. Please browse similar items
Polyfoil Polyester Lined Aluminum Foil, 8 x 8" Squares; 100/Box
Representative image only.
Polyfoil Mfr # F13200-0088 – Item # RK-06275-06
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyester-lined Aluminum
  • Width (in)8
  • Length (in)8
  • Width (cm)20.3
  • DescriptionPolyester Lined Aluminum Foil, 8 x 8\" Squares; 100/Box
More About this Item
Polyester laminated foil is resistant to punctures, tears and corrosion. Ideal for use with organic solvents and caustic solutions. Easy to apply and remove, folds easily, and resists kinking. Autoclavable. 100 per pack.