This item is now discontinued. Please browse similar items
Platinum Crucible Tongs, 9.8" L (250 mm)
Representative image only.
Mfr # TGS300 – Item # RK-17830-85
Prohibit contamination at high temperatures up to 2192°F (1200°C)
  • Protect your investment with platinum tongs that reduce possible damage and scratches to platinum crucible when hot
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPlatinum
  • DescriptionPlatinum Crucible Tongs, 9.8\" L (250 mm)
Key Features
  • Protect your investment with platinum tongs that reduce possible damage and scratches to platinum crucible when hot
More About this Item
Platinum offers oxidation resistance in air and oxygen; and corrosion resistance in acids and melted salts. Lids and crucibles sold separately.