This item is now discontinued. Please browse similar items
PIP 702 Nylon inspection gloves,standard weight, large, 12 pairs/pk
Representative image only.
PIP Mfr # 702 – Item # RK-86369-12
Low—lint nylon is an excellent choice for inspection of electronic components
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon
  • SizeLarge
  • Length (in)9
  • Length (cm)23
  • ColorWhite
  • CuffHemmed
  • SterileNo
  • DescriptionNylon inspection gloves,standard weight, large, 12 pairs/pk
More About this Item
Low-lint nylon inspection gloves are ideal for areas that are sensitive to lint particles often created by cotton gloves. These affordable inspection gloves are washable and reusable. Gloves are lightweight for extra sensitivity. Ambidextrous.
Nylon Inspection Gloves are lightweight for extra sensitivity.
Low-lint gloves are ideal for areas that are sensitive to lint particles often created by cotton gloves. Two-piece construction makes these unhemmed gloves comfortable enough for all day wear.