This item is now discontinued. Please browse similar items
PhotoDoc-It UVP 50 Imaging System; 115 VAC
Representative image only.
PhotoDoc-It Mfr # 97-0274-01 – Item # RK-97700-10
Compact system quickly prints and shoots brilliant color gel images with a high resolution camera
  • Instantly print color images on the photo printer included with the system
  • Save images to the memory card or connect to a computer via USB (not included)
  • Select systems include UV transilluminator
  • Ethidium bromide filter for fluorescent-stained gels
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Printer300 x 300 dpi resolution, 4\" x 6\" color prints; USB connectivity
  • Camera2.5\" LCD screen; USB connectivity
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionUVP 50 Imaging System; 115 VAC
  • Warranty1 year
1 Year Warranty
Key Features
  • Instantly print color images on the photo printer included with the system
  • Save images to the memory card or connect to a computer via USB (not included)
  • Select systems include UV transilluminator
  • Ethidium bromide filter for fluorescent-stained gels
  • 1 year warranty
More About this Item
System features a high-resolution, 16 megapixel digital color camera for capturing brilliant color images. Lightweight, durable hood enclosure provides a compact darkroom environment. The ethidium bromide emission filter, positioned inside the hood, is easily interchangeable for other filter types.
Models 97700-14 through -28 include a benchtop UV transilluminator to illuminate stained gels with the UV fluorescence. Color printer generates instant 4" x 6" (10.2 x 15.2 cm) color prints. Three-digit resettable combination lock secures the camera to the hood.